Reaction vessels, round bottom, with flat flange, Quickfit®

Leverandør: SciLabware
FR1LF FR250F FR500F FR2LF FR5LF FR10LF FR20LF FR3LF FR700F
201-8495EA 2175 NOK
201-8495 201-8511 201-8545 201-8521 201-8550 201-8478 201-8496 201-8527 BIBBFR700F
Reaction vessels, round bottom, with flat flange, Quickfit®
Reaksjonskolber Reaksjonskolber, kuleformet
PYREX®, borosilicate glass.

  • For use with Quickfit® flat flange lids and retaining clips
  • Excellent chemical resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

Wide neck allows easy incorporation of stirrer paddles and easy removal of solid residues.
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR