Transfection reagent diluent, 150 mM NaCl solution
Leverandør: Polyplus-transfection
201000002
201000002.EA 640 NOK
201000002.
Transfection reagent diluent, 150 mM NaCl solution
Transfeksjonsreagenser
This 150 mM NaCl solution is recommended as dilution reagent for DNA transfection with jetPEI®.
Since 1st of September 2021 Polyplus-Tranfection changed product part numbers. Products are remaining the same and the change is only related to the product part number. New product part numbers have symbol “New”. For duplicated products with symbol “telephone” please be informed that it’s the same product but with previous part number which we will only offer till the end of stock.
Sertifiseringer: Polyplus-transfection® has been an ISO 9001 accredited company since 2002. Since then this certification has been successfully renewed every three years.