Bilirubin conjugate, ditaurate, disodium salt, Calbiochem®
Leverandør: Merck Millipore Calbiochem
201102-100
201102-100EA 4250 NOK
201102-100
Bilirubin conjugate, ditaurate, disodium salt, Calbiochem®
Systemer for proteinkobling og -merking
Bilirubin conjugate, ditaurate is suitable as a direct bilirubin standard. It behaves similarly to purified bilirubin glucuronides from human bile in diazo tests.
Note: Unstable in solution.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Vert