Zincon sodium salt ≥75% (dye content), Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Warning


M-Clarity™ quality level = MQ100

201332-1G 201332-5G
201332-1GEA 680 NOK
201332-1G 201332-5G
Zincon sodium salt ≥75% (dye content), Sigma-Aldrich®
Zincon sodium salt

Zincon sodium salt has been used to locate zinc in plant tissues by light microscopy.


Zincon is a formazan dye, that serves as a metal indicator and is applicable in chelometric titration. It is used in spectrometric analysis. The metal-zincon complex formed shows a prominent absorption band in the visible region. Zincon is also used in solid support electrophoresis for staining zinc proteins.

Formel: C₂₀H₁₅N₄NaO₆S
Smeltepunkt: 250 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064385
CAS-nummer: 62625-22-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0411aa423a392b37b2aaf96f9c500c4d22d435d9&vid=0411aa423a392b37b2aaf96f9c500c4d22d435d9&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance (colour) Maroon to violet to purple
Appearance (form) Powder
Thin layer chromatogr. Corresponds to standard
Solubility (colour) Brown to very dark brown and red to very dark red and red-brown and very dark red-brown
Solubility (turbidity) Opaque
Solubility (method) 1 mg/ml, 0.1N sodium hydroxide
Carbon content 39.0 - 68.8%
Nitrogen content 9.1 - 15.2%
Infrared spectrum Conforms to structure
Wavelength (1) (UV) 489 - 495 nm
Molar absorbancy index (1) ≥10000
Solvent (UV) Methanol
Icp analysis Confirms sodium and sulfur components


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR