Chrysophenine 65% (dye content), Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

201901-50G
201901-50GEA 1190 NOK
201901-50G
Chrysophenine 65% (dye content), Sigma-Aldrich®
Chrysophenine

Chrysophenine, Dye content 65 % (Direct Yellow 12)

Formel: C₃₀H₂₈N₄O₈S₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007488
CAS-nummer: 2870-32-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e766fe866cd1d8327edee0532128e4b1a56a45a&vid=99fa66aa3f365e9bb80e917ab094f428b2a92f35&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR