Acid Yellow 25 40% (dye content), Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

201960-10G
201960-10GEA 745 NOK
201960-10G
Acid Yellow 25 40% (dye content), Sigma-Aldrich®
Acid Yellow 25

Acid Yellow 25, Dye content 40 %

Formel: C₂₃H₂N₅NaO₆S₂
MW: 549.55 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003121
CAS-nummer: 6359-85-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17542016063d10fd2b0c5127f5abc3044d1a84fc&vid=10794dba2887f9fcf8ae70d02b0e5c9e67d169b2&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR