Anti-IgG Donkey Polyclonal Antibody (CF™ 790)

Leverandør: Biotium
20344-50UL
20344-50ULEA 3875 NOK
20344-50UL
Anti-IgG Donkey Polyclonal Antibody (CF™ 790)
Antistoffer
Near-infrared CF™ dye antibodies are affinity-purified antibodies labeled with near-infrared fluorescent CF™750 or CF™770, two members of the novel CF™ dye series developed by Biotium for labeling proteins and other biomolecules. CF™ dyes are designed to offer advantages in brightness, photostability, and/or specificity compared to other commercial dyes, and are available in colors spanning the visible, far-red, and near-infrared spectra.

CF™680 has excitation/emission maxima at 681/698nm, and CF™680R has excitation/emission maxima at 680/701; these dyes are spectrally similar toMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG
Environmentally Preferable
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering CF® 790
Reaktivitet Rabbit
Vert Donkey
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoFluorescens Yes
Flowcytometri Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR