Jodhydrogensyre 57% stabilised analytical reagent

Leverandør: VWR Chemicals
Danger

Synonymer: Hydroiodic acid, Hydrogen iodide, Hydrogen iodide solution

20345.267EA 3950 NOK
20345.267
Jodhydrogensyre 57% stabilised analytical reagent
Jodhydrogensyre
Formel: HI
MW: 127.91 g/mol
Tetthet: 1.7 g/cm³ (20 °C)
MDL nummer: MFCD00011347
CAS-nummer: 10034-85-2
EINECS: 233-109-9
FN: 1787
ADR: 8 II

Bestill nå

Spesifikasjon Testresultater

Heavy metals (as Pb) Max. 10 ppm
Non-volatile residue Max. 0.5 %
Cl + Br (as Cl) Max. 0.03 %
SO4 (Sulphate) Max. 50 ppm
Fe (Iron) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR