Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Leverandør: Biotium
20353-1ML
20353-1MLEA 5750 NOK
20353-1ML
Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Antistoffer
R-PE belongs to the family of phycobiliproteins, which are derived from cyanobacteria and eukaryotic algae. R-PE has absorption maxima at 496, 546, and 565 nm respectively and emission at 578 nm, allowing it to be detected in the red channel by flow cytometry using 488 nm excitation. Because of its high extinction coefficient (~1,960,000) and high fluorescence quantum yield (~0.82), R-PE is by far the brightest dye for the 565 nm wavelength region.
Antibodies are supplied at 2 mg/mL in PBS pH 7.4 containing 50% glycerol, 2 mg/mL BSA (IgG- and protease-free) and 0.05% sodium azide. HighlyMore Product Information