Anti-6X His Mouse Monoclonal Antibody (CF770)

Leverandør: Biotium
20360
20360.EA 7450 NOK
20360.
Anti-6X His Mouse Monoclonal Antibody (CF770)
Antistoffer
Biotium offers a variety of anti-hapten and anti-tag fluorescent antibody conjugates. CF™ dyes offer advantages in brightness, photostability, and/or signal-to-noise ratio compared to other fluorescent dyes. Please see the CF™ Dye Quick Reference Table under the Datasheets tab to select the appropriate dye for your application.

Antibodies are supplied as affinity-purified IgG in PBS pH 7.4 containing 50% glycerol, 2 mg/mL BSA (IgG- and protease-free) and 0.05% sodium azide.

Type: Secondary
Antigen: 6X HIS TAG
Clonality: Monoclonal
Clone:
Conjugation: CF770
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Tag
Bestill nå

SPESIFIKASJONER

Antigensymbol HIS
Antigen-navn His Tag
Environmentally Preferable
Antistofftype Tag
Klonalitet Monoclonal
Konjugering CF® 770
Reaktivitet
Vert Mouse
Isotype IgG
ELISA Yes
Gelskifte Yes
Nøytralisering Yes
Konsentrasjon 1 mg/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR