Anti-IgG Goat Polyclonal Antibody (CF™ 660C)
Leverandør: Biotium
20368-50UL
20368-50ULEA 2425 NOK
20368-50UL
Anti-IgG Goat Polyclonal Antibody (CF™ 660C)
Antistoffer
CF™660C and CF™660R antibodies are affinity-purified antibodies labeled with far-red fluorescent CF™660C or CF™660R dyes, members of the novel CF™ dye series developed by Biotium for labeling proteins and other biomolecules. CF™ dyes are designed to offer advantages in brightness, photostability, and/or specificity compared to other commercial dyes, and are available in colors spanning the visible, far-red, and near-infrared spectra.
CF™660C is a far-red fluorescent dye with excitation/emission maxima at 667/685nm. CF™660C is several fold brighter and more photostable than the spectrallyMore Product Information