Glass wool, Supelco®
Leverandør: Merck
20383 20384 20409 20410 20411 21688-U
20383.EA 880 NOK
20383. 20384. 20409. 20410. 20411. 21688-U
Glass wool, Supelco®
Bomull og glassull
Wool plugs are often used in GC inlet liners to improve sample vaporization, and/or to keep non-volatile material from entering the column. They can also be used in packed GC columns, solvent desorption tubes, thermal desorption tubes, and purge traps to retain packed beds.
Silanised: For general purpose use
Pesticide Grade (Silanised): For applications that involve active analytes, such as organochlorine and/or organophosphorous pesticides.
Phosphoric acid treated: For acidic compounds such as barbiturates, free fatty acids, etc.
Non-treated: For special application where theMore Product Information