May Grunwald's eosin-methylene blue Cell Staining, Sigma-Aldrich®
Leverandør: SIGMA ALDRICH MICROSCOPY
Warning
MQ200
205435-25G
205435-25GEA 895 NOK
205435-25G
May Grunwald's eosin-methylene blue Cell Staining, Sigma-Aldrich®
May Grunwald's eosin-methylene blue
May-Grünwald stain is a Romanowsky neutraldye stain based on the combination ofmethylene blue azures as the basic dyecomponent and eosin as the acid dyecomponent.It is intended for use in staining blood films orbone marrow films. It is used routinely for diagnostic purposes in hematology.
  • May-Grünwald′s stain is used for the differential staining of cellular elements of blood
Bestill nå
Spesifikasjon Testresultater
Appearance (Colour) Green to Black
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
UV Absorbance Ratio
A650/A523
≥ 1.45