Bromoxylenol blue, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY


MQ200

205478-10G
205478-10GEA 1580 NOK
205478-10G
Bromoxylenol blue, Sigma-Aldrich®
Bromoxylenol blue
Formel: C₂₃H₂₀Br₂O₅S
MDL nummer: MFCD00005873
CAS-nummer: 40070-59-5
EINECS: 254-780-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b4e907133ae46f99b834c6b9ed9b828e78315ec&vid=70fb49aa0affab7aa59f9f52d83ef84151b28e03&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR