Caffeic acid, Millipore®

Leverandør: Merck Millipore Calbiochem

Synonymer: 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid

205546-500
205546-500EA 1875 NOK
205546-500
Caffeic acid, Millipore®
Caffeic acid
Formel: C₉H₈O₄
MW: 180,16 g/mol
Kokepunkt: 366 °C (1013 hPa)
Smeltepunkt: 234…237 °C
MDL nummer: MFCD00004392
CAS-nummer: 331-39-5
EINECS: 206-361-2
Merck Indeks: 12,01673

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR