Perchloric acid 70% ≥70%, GPR RECTAPUR®

Leverandør: VWR Chemicals
Danger

20587.296EA 3125 NOK
20587.296
Perchloric acid 70% ≥70%, GPR RECTAPUR®
Perchloric acid
Formel: HClO₄
MW: 100.46 g/mol
Kokepunkt: 160…181 °C (1013 hPa)
Smeltepunkt: –112,2 °C
Tetthet: 1.53…1.768 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011325
CAS-nummer: 7601-90-3
EINECS: 231-512-4
FN: 1873
ADR: 5.1 (8) I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay 69 - 72 %
Ignition residue (SO4) Max. 0.02 %
Cl (Chloride) Max. 25 ppm
Fe (Iron) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR