iso-Amylalkohol ≥99.0%, AnalaR NORMAPUR® analytical reagent

Leverandør: VWR Chemicals
Warning

Synonymer: Isopentyl alcohol, iso-Pentanol, 3-Methyl-1-butanol, 3-Methylbutanol

20798.295EA 1325 NOK
20798.295
iso-Amylalkohol ≥99.0%, AnalaR NORMAPUR® analytical reagent
iso-Amylalkohol
Formel: (CH₃)₂CHCH₂CH₂OH
MW: 88.15 g/mol
Kokepunkt: 130
Smeltepunkt: –117 °C
Tetthet: 0.812 g/cm³ (20 °C)
Flash punkt: 42
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002934
CAS-nummer: 123-51-3
EINECS: 204-633-5
FN: 1105
ADR: 3 III
Merck Indeks: 13,05214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd7b89cc1890b5d4b2083195c30c0d1047c32e9a&vid=170c60e26c7b4edab6e0686f91d04e676da16e6d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (calculated as mixture of isomers) Min. 99.0 %
Acidity Max. 0.008 meq/g
Alkalinity Max. 0.0008 meq/g
Boiling point 131 - 132.5 °C
Colouration Max. 10 APHA
Density (20/4) 0.808 - 0.812
Acidity and esters (as C7H14O2) Max. 0.17 %
Evaporation residue Max. 10 ppm
Furfuraldehyde Max. 1 ppm
Pentan-1-ol Max. 0.5 %
Peroxides (as H2O2) Max. 10 ppm
Water Max. 0.2 %
Al (Aluminium) Max. 0.5 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.02 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.1 ppm
Sn (Tin) Max. 0.1 ppm
Zn (Zinc) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR