2-Propanol, Wasser_0,01 99,8, AnalaR NORMAPUR® zA

Leverandør: VWR Chemicals
Danger

Synonymer: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

20838.294EA 565 NOK
20838.294
2-Propanol, Wasser_0,01 99,8, AnalaR NORMAPUR® zA
2-Propanol
Formel: (CH₃)₂CHOH
MW: 60.1 g/mol
Kokepunkt: 82
Smeltepunkt: –89 °C
Tetthet: 0.786 g/cm³ (20 °C)
Flash punkt: 12
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011674
CAS-nummer: 67-63-0
EINECS: 200-661-7
FN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Indeks: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colourless liquid
Colour value Max. 10 APHA
Assay Min. 99.8 %
Aldehydes Max. 0.0002 %
Formaldehyde Max. 0.0002 %
Ketones (as C3H6O) Max. 0.0050 %
Methanol Max. 0.1 %
Residue on evaporation Max. 0.0010 %
Water (K.F.) Max. 0.0100 %
Formaldehyde-sulphuric colouration Max. 60.0 APHA
Acidity Max. 0.0020 %
Al (Aluminium) Max. 0.5 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.1 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.01 ppm
Fe (Iron) Max. 0.1 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.01 ppm
Ni (Nickel) Max. 0.01 ppm
Pb (Lead) Max. 0.01 ppm
Sn (Tin) Max. 0.1 ppm
Zn (Zinc) Max. 0.01 ppm

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR