Metanol bio-based 99,5, GPR RECTAPUR®

Leverandør: VWR Chemicals
Danger

Synonymer: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

20844.292EA 480 NOK
20844.292 20844.326 20844.463 20844.554
Metanol bio-based 99,5, GPR RECTAPUR®
Metanol
Formel: CH₃OH
MW: 32.04 g/mol
Kokepunkt: 64.6 °C (1013 hPa)
Smeltepunkt: –98 °C
Tetthet: 0.7918 g/cm³ (20 °C)
Flash punkt: 11 °C (closed cup)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004595
CAS-nummer: 67-56-1
EINECS: 200-659-6
FN: 1230
ADR: 3 (6.1) II
REACH: 01-2119433307-44
Merck Indeks: 13,05984

Bestill nå

Spesifikasjon Testresultater

Assay (GC) min. 99.5%
Appearance clear colourless liquid
IR Spectrum Passes test
Acidity (meq/g) max. 0.0002
Boiling point 64 - 65°C
Colouration (APHA) max. 10
Acetone max. 10 ppm
Evaporation residue max. 50 ppm
Water max. 0.2%

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR