Human AB pooled serum (male), French origin

Leverandør: SEQENS
21001PM-100ML
21001PM-100MLEA 4750 NOK
21001PM-100ML
Human AB pooled serum (male), French origin
Serum
SEQENS IVD offers Human AB Pooled Serum (male), collected from French healthy AB male donors.

  • Dry extract: 70 to 90 g/L
  • Total proteins: 60 to 80 g/L
  • Filtered through a 0,2 µm membrane
  • Shelf life: 3 years

Each donor unit used for the manufacture of this product has been tested and found negative or non-reactive (CE marked test methods) for anti-HIV 1+2 and anti-HCV antibodies, HBs antigen, HIV 1 and HCV RNA and Syphilitic serology.

Complete donor traceability on file.

NB: Only for research or manufacture of in vitro diagnostic reagents. Not forMore Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR