Acid blue 80 40% (dye content), Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Warning

Synonymer: Alizarine Fast Blue R, Nylosan Blue C-L, Endanil Blue B

210323-100G
210323-100GEA 1980 NOK
210323-100G
Acid blue 80 40% (dye content), Sigma-Aldrich®
Acid blue 80

Acid Blue 80, Dye content 40 %

Formel: C₃₂H₂₈N₂Na₂O₈S₂
MW: 678.682 g/mol
Smeltepunkt: >300 °C (lit.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001192
CAS-nummer: 4474-24-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50c94cb0a414b99f0ef91af441c7822a873edb71&vid=d2d08886cbd16c2b1d050c717b19660ec1be9078&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR