Coomassie violet 50% (dye content) R200, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

Synonymer: C.I. 42650, Acid violet 17

210579-50G
210579-50GEA 963 NOK
210579-50G
Coomassie violet 50% (dye content) R200, Sigma-Aldrich®
Coomassie violet

Coomassie Violet R200, Dye content 50 % (Acid violet 17)

Formel: C₄₁H₄₇N₃NaO₆S₂
MW: 761.92 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012120
CAS-nummer: 4129-84-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6fdac8931adf5227f57c50e74ae758538e7ecd1&vid=79aac65bf34d5d41e7db8b3903d8e4b81242b2ef&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR