Acid red 1 60% (dye content), Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

Synonymer: Red 2G, C.I. 18050, Amido Naphthol Red G, Sodium 5-acetamido-4-hydroxy-3-(phenyldiazenyl)naphthalene-2,7-disulphonate, 5-Acetamido-4-hydroxy-3-phenylazo-2,7-naphthalene-disulphonic acid disodium salt, Pontacyl Carmine, Azophloxine

M-Clarity™ quality level = MQ100

210633-100G 210633-25G
210633-100GEA 1900 NOK
210633-100G 210633-25G
Acid red 1 60% (dye content), Sigma-Aldrich®
Acid red 1
Formel: C₁₈H₁₅N₃O₈S₂
Lagringstemperatur: Room Temperature
MDL nummer: MFCD00003954
CAS-nummer: 3734-67-6
EINECS: 223-098-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7c7165c3e0511e1301f8893984d66075e9c8f68&vid=a7c7165c3e0511e1301f8893984d66075e9c8f68&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance (Color) Red to Purple
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Wavelength 503 - 509 nm
Extinction Coefficient ≥17000
Extinction Coefficient c = 0.02g/l; Water ≥17000
Carbon 25.3 - 59.1%
Nitrogen 4.5 - 12.0%

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR