Acid blue 25 45% (dye content), Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Warning

Synonymer: Sodium 1-amino-9,10-dioxo-4-phenylaminoanthracene-2-sulphonate, C.I. 62055, sodium 1-amino-9,10-dioxo-4-(phenylamino)-9,10-dihydroanthracene-2-sulphonate

210684-25G
210684-25GEA 1720 NOK
210684-25G
Acid blue 25 45% (dye content), Sigma-Aldrich®
Acid blue 25

Acid Blue 25, Dye content 45 %

Formel: C₂₀H₁₃N₂O₅SNa
MW: 416.4 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001214
CAS-nummer: 6408-78-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a8550533ef407d176554a90c832e243577a9663&vid=1c27682d3863e5d287ca00b123fc3c1a85f47e89&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR