Fenolftalein 1% in ethanol 50% pH-indicator

Leverandør: VWR Chemicals
Danger

Synonymer: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

210894REA 885 NOK
210894R
Fenolftalein 1% in ethanol 50% pH-indicator
Fenolftalein

1% solution in 50% ethanol.


  • 1% solution in 50% ethanol
  • Transition range: pH 8.2 to 9.8

Formel: C₂₀H₁₄O₄
MW: 318.33 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005913
CAS-nummer: 77-09-8
EINECS: 201-004-7
FN: 1170
ADR: 3 III
Merck Indeks: 14,07243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Application test Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR