VWR®, Cryoboxes, for Light-Sensitive Specimens
Leverandør: VWR Collection
211-0255EA 280 NOK
211-0255
VWR®, Cryoboxes, for Light-Sensitive Specimens
Bokser Kryobokser
PP, black.
  • Reinforced seal, triple-hinges and locking clasp keeps samples secure while the box is closed
  • Each box features an alphanumeric grid on the exterior and interior and stacking rings
  • Matt surface suitable for printing and labelling
Ideal for light-sensitive specimens, this all-black storage box is suitable for use with 0,5/1,5/2,0 ml microcentrifuge tubes, as well as most other brands of cryogenic vials. These boxes stand a few millimetres taller than other available types of storage boxes and can accommodate several sizes of cryogenic vials,More Product Information