Racks for micro sample test tubes, PP, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
F188410000
211-7420EA 1200 NOK
211-7420
Racks for micro sample test tubes, PP, SP Bel-Art
Stativer for rør Tube Racks
PP, white.
  • Very good chemical resistance
  • Very good temperature resistance
These test tube racks stack to conserve space, even when filled with tubes.