Cerium(IV) oxide, hydrated

Leverandør: Thermo Fisher Scientific
Danger

21120.18 21120.30
21120.18EA 950 NOK
21120.18 21120.30
Cerium(IV) oxide, hydrated
Cerium(IV) oxide, hydrated

CAS No.: 12014-56-1 Molecular Formula: CeO2.xH2O Formula Weight: 172.12(anhy) Physical Form: Lump/Powder Appearance: Yellow to pale brown Odour: Odourless UN No.: UN3262 MDL No.: MFCD00010927

Formel: CeO₂.xH₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010927
CAS-nummer: 12014-56-1
EINECS: 234-599-7
ADR: 8,III

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR