Calcium ionophore I ≥99.0%, Selectophore™, function tested ionophore, Supelco®

Leverandør: Merck

Synonymer: ETH 1001, (−)-(R,R)-N,N′-Bis-[11-(ethoxycarbonyl)undecyl]-N,N′,4,5-tetramethyl-3,6-dioxaoctane-diamide, Diethyl N,N′-[(4R,5R)-4,5-dimethyl-1,8-dioxo-3,6-dioxaoctamethylene]bis(12-methylaminododecanoate)

M-Clarity™ quality level = MQ100

21192-250MG-F 21192-50MG-F
21192-250MG-FEA 19925 NOK
21192-250MG-F 21192-50MG-F
Calcium ionophore I ≥99.0%, Selectophore™, function tested ionophore, Supelco®
Calcium ionophore I

Used as ionophore in membrane electrodes for the determination of Ca2+-activities.


Calcium ionophore I (ETH 1001) was used in an experimental study, conducted to determine the intracellular and extracellular changes of calcium ions in hypoxia and ischemia in rat brain.


Useful for transport studies in biological membranes.


Forpakning: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₃₈H₇₂N₂O₈
MDL nummer: MFCD00042943
CAS-nummer: 58801-34-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d23cbd64b90e5cce48628d19509ed72912ca345&vid=bfa34e919e15d87db4d78e7b5b3e3ed6cf59f815&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR