Calcium ionophore II for ion selective electrodes, Selectophore™, Supelco®
Leverandør: Merck
21193-250MG-F 21193-50MG-F
21193-250MG-FEA 8075 NOK
21193-250MG-F 21193-50MG-F
Calcium ionophore II for ion selective electrodes, Selectophore™, Supelco®
Elektrolytter Løsninger, elektroder
Lipophilic ionophore for Ca²⁺ selective electrodes with detection limit in the sub-nanomolar range.
Extremely efficient carrier of Ca²⁺ -ions; induces a selectivity in membranes for Ca²⁺ over Mg²⁺ by a factor of about 10⁴. Calcium ionophore II (ETH 129) was used in preparing calcium selective minielectrodes which are used in determination of dissociation constant (Kd).
Forpakning: Packed in bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79dce6fa0178eb2f82e1641265ee048bc95a06bb&vid=79dce6fa0178eb2f82e1641265ee048bc95a06bb&prid=web&clid=SAPDAM
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
M-Clarity quality level MQ100