VWR®, Test Tubes Racks
Leverandør: VWR Collection
212-0409EA 290 NOK
212-0409 212-0422 212-0421 212-0420 212-0419 212-0418 212-0417 212-0416 212-0415 212-0414 212-0413 212-0412 212-0411 212-0425 212-0407 212-0393 212-0406 212-0405 212-0404 212-0403 212-0402 212-0401 212-0400 212-0399 212-0398 212-0397 212-0396 212-0382 212-0394 212-0410 212-0392 212-0391 212-0390 212-0389 212-0388 212-0387 212-0386 212-0385 212-0384 212-0383 212-0395 212-0423 212-0430 212-0429 212-0428 212-0427 212-0426 212-0408 212-0424
VWR®, Test Tubes Racks
Stativer for rør Tube Racks
Nylon, glass fibre reinforced.
  • Racks are single-piece moulded
  • Good heat and chemical resistance
  • Three tiers with contents always clearly visible
  • Large labelling areas on the side surfaces