VWR®, Test Tubes/Centrifuge Tubes Racks
Leverandør: VWR Collection
212-0705EA 545 NOK
212-0705 212-0704
VWR®, Test Tubes/Centrifuge Tubes Racks
Stativer for rør Tube Racks
PP, PS or steel wire with epoxy-coating.
  • Very good chemical and thermal resistance, autoclavable
  • PP racks are interlocking
  • Robust, firm
Ideal for accommodating a single 50 ml tube with a round or conical bottom.
Leveringsinformasjon: The PP interlocking racks are supplied in five assorted colours, blue, green, yellow, pink and orange.