VWR®, Racks for Test Tubes
Leverandør: VWR Collection
212-0713EA 430 NOK
212-0713 212-0714 212-0715 212-0716 212-0717 212-0718 212-0719 212-0720 212-0721 212-0722 212-0723 212-0724 212-0725
VWR®, Racks for Test Tubes
Stativer for rør Tube Racks
These test tube racks are either made from rust-proof, electrolytically polished stainless steel or from polyamide coated steel.
  • Very good chemical and temperature resistance
  • Polyamide coated racks are heat resistant up to 80 °C
  • Robust and stable, easy to clean
  • Hole size 18×18 mm