Beger, trekantede, Tricorn™
Leverandør: Simport Scientific
B700-100 B700-1L B700-250 B700-400 B700-800 B700-50
212-9302EA 820 NOK
212-9302 212-9306 212-9303 212-9304 212-9305 212-9301
Beger, trekantede, Tricorn™
Beger
These Tricorn™ beakers are made of polypropylene, provide three dripless pouring spouts that offers more security.
  • Autoclavable
  • Tough and unbreakable
Suitable for use with commonly used acids, alkalis and solvents.
Forpakning: All sizes are available 100 per case.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904