Direct Blue 71 50% Dye, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

212407-50G
212407-50GEA 963 NOK
212407-50G
Direct Blue 71 50% Dye, Sigma-Aldrich®
Direct Blue 71

Direct Blue 71, Dye content 50 %

Formel: C₄H₂₃N₇O₁₃S₄
MW: 1029.87 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003971
CAS-nummer: 4399-55-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7807d46afe3f377e536dbde183b6ce6f18471d5d&vid=7b3e5f99367285b906cfbd5ce23719557e7da672&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR