di-Ammoniumhydrogencitrat ≥97%, GPR RECTAPUR®

Leverandør: VWR Chemicals
Warning

Synonymer: Citric acid diammonium salt, Diammonium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate, Ammonium citrate dibasic

21245.361EA 3925 NOK
21245.361
di-Ammoniumhydrogencitrat ≥97%, GPR RECTAPUR®
di-Ammoniumhydrogencitrat
Formel: HOC(CO₂H)(CH₂CO₂NH₄)₂
MW: 226.19 g/mol
Tetthet: 1.483 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013068
CAS-nummer: 3012-65-5
EINECS: 221-146-3
Merck Indeks: 13,00513

Bestill nå

Spesifikasjon Testresultater

Assay Min. 97 %
Heavy metals (as Pb) Max. 20 ppm
Ignition residue (SO4) Max. 0.1 %
Fe (Iron) Max. 20 ppm

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR