VWR® Beakers, Low Form, PMP (TPX)
Leverandør: VWR Collection
213-0510EA 160 NOK
213-0510 213-0482 213-0483 213-0484 213-0485 213-0486 213-0487 213-0488
VWR® Beakers, Low Form, PMP (TPX)
Beger
Made of PMP (TPX), crystal-clear.
  • Excellent chemical resistance
  • Printed graduations
  • Withstand temperatures of 0 to 121 °C when used continuously (up to 170 °C for short periods)
  • Resin meets US FDA 21 CFR requirements