Ammoniumdihydrogenortofosfat ≥97%, TECHNICAL

Leverandør: VWR Chemicals

21302.368EA 1950 NOK
21302.368
Ammoniumdihydrogenortofosfat ≥97%, TECHNICAL
Ammoniumdihydrogenortofosfat
Formel: NH₄H₂PO₄
Smeltepunkt: 190 °C
Tetthet: 1,8 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003396
CAS-nummer: 7722-76-1
EINECS: 231-764-5
REACH: 01-2119488166-29
Merck Indeks: 13,00547

Bestill nå

Spesifikasjon Testresultater

Assay Min. 97 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR