Ammoniumsulfat ≥99%, GPR RECTAPUR®

Leverandør: VWR Chemicals

21332.293EA 480 NOK
21332.293 21332.362 21332.460
Ammoniumsulfat ≥99%, GPR RECTAPUR®
Ammonium sulphate
Formel: (NH₄)₂SO₄
Smeltepunkt: 336…339 °C
Tetthet: 1,78 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003391
CAS-nummer: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Indeks: 13,00559

Bestill nå

Spesifikasjon Testresultater

Assay Min. 99 %
Heavy metals (as Pb) Max. 10 ppm
Ignition residue (SO4) Max. 0.02 %
Cl (Chloride) Max. 20 ppm
NO3 (Nitrate) Max. 50 ppm
Fe (Iron) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR