VWR®, Erlenmeyer Flasks with Four Baffles
Leverandør: VWR Collection
214-0436EA 280 NOK
214-0436 214-0437 214-0438 214-0439
VWR®, Erlenmeyer Flasks with Four Baffles
Kolber Erlenmeyerkolber
Borosilicate 3.3 glass, with 4 baffles, GL 45 thread and blue GL 45 membrane screw cap.
  • High chemical and thermal resistance
  • With 4 equidistant baffles providing consistent agitation when used with orbital shakers
  • Ideal for bacterial culture production
  • Complete with blue GL 45 membrane screw cap made from PP
  • Autoclavable at 121 °C
The 4 equidistant baffles of these flasks are providing consistent agitation when used with orbital shakers. The VWR® baffled flasks are delivered with a GL 45 membrane screw cap with 0,2 µm PTFE membrane to ensures anMore Product Information