VWR®, PETG Erlenmeyer Shaker Flask
Leverandør: VWR Collection
214-0440EA 1550 NOK
214-0440 214-0441 214-0443 214-0444 214-0442 214-0445 214-0454 214-0455
VWR®, PETG Erlenmeyer Shaker Flask
Kolber Erlenmeyerkolber
VWR® crystal-clear PETG Erlenmeyer shaker flasks are robust and break resistant, increasing personal safety. They are ideal for shaker culture applications and storage.
  • Temperature range: −60 to +60 °C
  • Unlike glass, flasks will not break when dropped
  • Moulded-in graduations for accuracy
  • Vent cap option for continuous gas exchange while ensuring sterility and preventing leakage
  • Non cytotoxic, DNase-/RNase-free and non pyrogenic
  • Sterilised by irradiation SAL 10⁻⁶ (ISO 11137)
  • Shelf life: 3 years after the month of production
Manufactured in a Class 100,000More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat ISO 14001/Certificate No.: 04923E00243R0M