VWR®, PC Erlenmeyer Shaker Flasks
Leverandør: VWR Collection
214-0446EA 1600 NOK
214-0446 214-0447 214-0448 214-0449 214-0450 214-0451 214-0452 214-0453 214-0476 214-0477 214-0478 214-0479 214-0480 214-0481
VWR®, PC Erlenmeyer Shaker Flasks
Kolber Erlenmeyerkolber
VWR® PC erlenmeyer shaker flasks provide excellent optical clarity and mechanical strength. They are ideal for shaker culture applications and storage.
  • Temperature range: –80 to +121 °C
  • Unlike glass, flasks will not break when dropped
  • Molded-in graduations for accuracy
  • Vent cap option for continuous gas exchange while ensuring sterility and preventing leakage
  • Non-cytotoxic, DNase/RNase free and non-pyrogenic
  • Sterilized by irradiation SAL10-6 (ISO11137)
  • Shelf life: 3 years after month of production
Manufactured in a class 100,000 room environment underMore Product Information