Alizarin Cyanin Green F ≥60 % (dye content), Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

Synonymer: acid Green 25, D and C Green No. 5, Acid Green 25, Alizarin Cyanin Green F

214566-50G
214566-50GEA 894 NOK
214566-50G
Alizarin Cyanin Green F ≥60 % (dye content), Sigma-Aldrich®
Alizarin cyanin green F

Acid Green 25, Dye content ≥60 %

Formel: C₂₈H₂₂N₂O₈S₂
MW: 622.57 g/mol
Smeltepunkt: 235…238 °C (lit.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001193
CAS-nummer: 4403-90-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1af5c870327f6bf91a268acf3c81ef8512a9b97&vid=8df77a13ba3b3c9a2e68fc8ccf4f41289f87db12&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR