Antimon(III)klorid 99,0, AnalaR NORMAPUR® zA

Leverandør: VWR Chemicals
Danger

Synonymer: Antimony trichloride

21470.186EA 1200 NOK
21470.186
Antimon(III)klorid 99,0, AnalaR NORMAPUR® zA
Antimon(III)klorid
Formel: SbCl₃
MW: 228.12 g/mol
Kokepunkt: 223.5 °C (1013 hPa)
Smeltepunkt: 73
Tetthet: 3.14 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011212
CAS-nummer: 10025-91-9
EINECS: 233-047-2
FN: 1733
ADR: 8 II
Merck Indeks: 13,00713

Bestill nå

Spesifikasjon Testresultater

Assay Min. 99.0 %
Insolubility in chloroform Max. 0.01 %
Insolubility in HCl Max. 50 ppm
SO4 (Sulphate) Max. 50 ppm
Ca (Calcium) Max. 50 ppm
Cu (Copper) Max. 5 ppm
Fe (Iron) Max. 20 ppm
K (Potassium) Max. 0.01 %
Na (Sodium) Max. 50 ppm
Pb (Lead) Max. 30 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR