Antimony(III) potassium oxitartrate trihydrate ≥99%, GPR RECTAPUR®

Leverandør: VWR Chemicals
Warning

Synonymer: (R,R)-(+)-Antimony potassium tartrate trihydrate, (+)-Antimony potassium L-tartrate trihydrate, Tartar emetic, Antimony potassium L(+)-tartrate trihydrate, L(+)-Tartaric acid antimony potassium salt trihydrate, Potassium Antimony (III) Oxide Tartrate Trihydrate, (R,R)-(+)-Tartaric acid antimony potassium salt trihydrate, L(+)-Antimony potassium tartrate trihydrate, Potassium antimony(III) oxide tartrate trihydrate, Antimony potassium (+) tartrate trihydrate, Potassium antimony tartrate trihydrate, Potassium antimonyl tartrate sesquihydrate, Antimony potassium tartrate trihydrate, Potassium antimonyl tartrate trihydrate

21486.237EA 685 NOK
21486.237
Antimony(III) potassium oxitartrate trihydrate ≥99%, GPR RECTAPUR®
Antimony(III) potassium oxitartrate trihydrate
Formel: C₈H₄K₂O₁₂Sb₂·3H₂O
Smeltepunkt: 100 °C
Tetthet: 2,6 g/cm³ (20 °C)
MDL nummer: MFCD00167056
CAS-nummer: 28300-74-5
EINECS: 229-436-1
FN: 1551
ADR: 6.1,III
Merck Indeks: 13,00706

Bestill nå

Spesifikasjon Testresultater

Assay Min. 99 %
Cl (Chloride) Max. 100 ppm
As (Arsenic) Max. 0.025 %
Fe (Iron) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR