Centrifuge bottles, PP/PE, with screw cap
Leverandør: SEMADENI
2139
215-0224EA 685 NOK
215-0224
Centrifuge bottles, PP/PE, with screw cap
Flasker Sentrifugeflasker
PPCO, translucent with PE screw cap.
  • Autoclavable
  • Excellent chemical resistance
  • Withstands forces up to 27500×g for 250 ml, 13500×g for 500 ml and 7100×g for 1000 ml