Wash bottles, wide neck, for microbiological staining solutions, Azlon®
Leverandør: SciLabware
WGW0255V
215-0595EA 775 NOK
215-0595
Wash bottles, wide neck, for microbiological staining solutions, Azlon®
Flasker Spruteflasker
LDPE, med PP skrulokk.
  • Set of five 250 ml venting wash bottles with DripLok® anti-drip valve
  • Printed for Lugols Iodine, Crystal Violet, Gram Colour Decolourising, Gram Iodine and Safranine Solutions
  • Printed GHS compliant hazard information where relevant and instructions
Sertifiseringer: Azlon® wash bottles meet the material requirements of the U.S. Food and Drug Administration as specified in 21 CFR 177.1520.