Flasker, smal munn, HDPE, med skrulokk, for miljøprøver, rund, Nalgene®
Leverandør: Thermo Fisher Scientific
2089-0002 2089-0004 2089-0008 2089-0016 2089-0032
215-0998EA 1475 NOK
215-0998 215-1027 215-1028 215-1029 215-1030
Flasker, smal munn, HDPE, med skrulokk, for miljøprøver, rund, Nalgene®
Flasker Plastflasker
HDPE, naturfarget med PP-skrulokk.
  • God kjemisk resistens
  • Varmebestandig fra −100 til +120 °C
  • Lekkasjesikker
Narrow neck economy bottles are lightweight versions of the original Nalgene® Lab Quality Bottle, suitable for lighter-duty, general purpose applications.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact,Product Transparency and Disclosure Low Manufacturing Impact,Product Transparency and Disclosure
Miljøsertifikat NAR Certificate of Retirement, ACT Label NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603, ACT Label