Laboratorieflasker, vid hals, klar, med GLS 80® gjenger, DURAN®
Leverandør: DWK Life Sciences
1178392 1178424 1178425 1178426 1178427 1178428 218606904 218603607
215-3094EA 2600 NOK
215-3094 215-3095 215-3096 215-3097 215-3098 215-3099 215-2849 215-2957
Laboratorieflasker, vid hals, klar, med GLS 80® gjenger, DURAN®
Flasker Glassflasker
DURAN®, borosilicate glass 3.3, clear, without screw caps and pouring rings.
  • Easy-to-read scale and large labelling field for easy marking, in fired-on, highly durable white ceramic
  • Very good chemical resistance, high temperature resistance
  • Special thread means opening takes less than a turn
  • Service temperature limit of standard blue closure and pouring ring (must be ordered separately): 140 °C
  • Glass Type I according to USP, EP and JP
Round, with graduations and specially developed GLS 80 thread. Opening and closing the bottle is easy with quickMore Product Information