Laboratory bottles, stainless steel, with thread, without screw cap, DURAN®
Leverandør: DWK Life Sciences
299016006
215-3262EA 11150 NOK
215-3262
Laboratory bottles, stainless steel, with thread, without screw cap, DURAN®
Flasker Metallflasker
Manufactured from corrosion resistant AISI Type 316 L (1.4404) stainless steel, and hygienically constructed with all welded seams.
  • Hazardous materials laboratory container for storage of liquids, intermediates, or solid product
  • Storage of precious materials, such as high purity fine chemicals, pharmaceutical or cosmetic products
  • Storage of light sensitive materials
The unbreakable steel bottle is ideal for storage applications where glass is not applicable, due to the risk of breakage or chemical incompatibility. The bottle has a smooth inner surfaceMore Product Information